CAS RN 6314-42-7
IUPAC Name:
1-benzothiophene-2-carboxamide
Synonyms:
1-benzothiophene-2-carboxamide
Molecular Weight (g/mol):
177.22
Molecular Formula:
C9H7NOS
InChi Key:
GYSCBCSGKXNZRH-UHFFFAOYSA-N
SMILES:
NC(=O)C1=CC2=CC=CC=C2S1
1
–
1
of
1
results
1
Promotions Available
Benzo[b]thiophene-2-carboxamide, 97%
Benzo[b]thiophene-2-carboxamide, 97%
CAS: 6314-42-7 Molecular Formula: C9H7NOS Molecular Weight (g/mol): 177.221 MDL Number: MFCD00052505 InChI Key: GYSCBCSGKXNZRH-UHFFFAOYSA-N Synonym: benzo b thiophene-2-carboxamide,thianaphthene-2-carboxamide,benzothiophene carboxamide,2-benzo b thiophenecarboxamide,# PubChem CID: 237073 IUPAC Name: 1-benzothiophene-2-carboxamide SMILES: C1=CC=C2C(=C1)C=C(S2)C(=O)N