missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]thiophene-2-carbonyl Chloride 98.0+%, TCI America™

Catalog No. B46375G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B46375G 5 g
B46371G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B46375G Supplier TCI America Supplier No. B46375G

Chemical Identifiers

CAS 39827-11-7
Molecular Formula C9H5ClOS
Molecular Weight (g/mol) 196.648
MDL Number MFCD00051635
InChI Key DNGLRCHMGDDHNC-UHFFFAOYSA-N
Synonym benzo b thiophene-2-carbonyl chloride, thianaphthene-2-carbonyl chloride, benzo b thiophene-2-carbonylchloride, acmc-20alq1, 2-benzthiophenecarbonylchloride, 2-benzothiophenecarbonyl chloride, benzothiophene-2-carbonyl chloride, 2-benzo b thiophenecarbonyl chloride, 2-chlorocarbonyl-1-benzothiophene
PubChem CID 142384
IUPAC Name 1-benzothiophene-2-carbonyl chloride
SMILES C1=CC=C2C(=C1)C=C(S2)C(=O)Cl

Specifications

Melting Point 84°C
Color White-Yellow
Boiling Point 176°C
Quantity 5 g
UN Number 3261
Formula Weight 196.65
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]thiophene-2-carbonyl Chloride