accessibility menu, dialog, popup

UserName

CAS RN 39827-11-7

ClOS

CAS RN 39827-11-7

IUPAC Name: 1-benzothiophene-2-carbonyl chloride
Synonyms: 1-benzothiophene-2-carbonyl chloride
Molecular Weight (g/mol): 196.65
Molecular Formula: C9H5ClOS
InChi Key: DNGLRCHMGDDHNC-UHFFFAOYSA-N
SMILES: ClC(=O)C1=CC2=CC=CC=C2S1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results