missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]naphtho[1,2-d]thiophene 98.0+%, TCI America™
Supplier: TCI America B49515G
Specifications
| Benzo[b]naphtho[1,2-d]thiophene | |
| 103°C | |
| 220°C | |
| MFCD00215942 | |
| XZUMOEVHCZXMTR-UHFFFAOYSA-N | |
| naphtho[2,1-b][1]benzothiole | |
| 9151 | |
| ≥98.0% (GC) |
| 205-43-6 | |
| White-Yellow | |
| C16H10S | |
| 5 g | |
| C1=CC=C2C(=C1)C=CC3=C2C4=CC=CC=C4S3 | |
| 234.316 | |
| 234.32 | |
| Crystalline Powder |
Chemical Identifiers
| 205-43-6 | |
| 234.316 | |
| XZUMOEVHCZXMTR-UHFFFAOYSA-N | |
| naphtho[2,1-b][1]benzothiole |
| C16H10S | |
| MFCD00215942 | |
| 9151 | |
| C1=CC=C2C(=C1)C=CC3=C2C4=CC=CC=C4S3 |
Safety and Handling
RTECSNumber : DI2340000
TSCA : No
Recommended Storage : Refrigerator