missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]naphtho[1,2-d]thiophene 98.0+%, TCI America™

Catalog No. B49511G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B49511G 1 g
B49515G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B49511G Supplier TCI America Supplier No. B49511G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 205-43-6
Molecular Formula C16H10S
Molecular Weight (g/mol) 234.316
MDL Number MFCD00215942
InChI Key XZUMOEVHCZXMTR-UHFFFAOYSA-N
PubChem CID 9151
IUPAC Name naphtho[2,1-b][1]benzothiole
SMILES C1=CC=C2C(=C1)C=CC3=C2C4=CC=CC=C4S3

Specifications

Melting Point 103°C
Color White-Yellow
Boiling Point 220°C
Quantity 1 g
Formula Weight 234.32
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]naphtho[1,2-d]thiophene
RTECSNumber DI2340000
TSCA No
Recommended Storage Refrigerator