missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan, 97+%

Catalog No. AAA1522509
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1522509 10 g
AAA1522518 50 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1522509 Supplier Thermo Scientific Chemicals Supplier No. A1522509
Get Up to $1,800 in Chemicals with Purchase
View all promotions

CAS: 271-89-6 | C8H6O | 118.14 g/mol

Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzo[b]furan is used in the manufacture of coumarone-indene resin. It has been used to make floor tiles and other products. It is also used as a coating on grapefruit, lemons, lirnes, oranges, tangelos and tangerines and in the production of COITosion-resistant paints and varnishes, in water-resistant coatings on paper products and fabrics and as adhesives in food containers.

Solubility
Not miscible in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents and heat.

Chemical Identifiers

CAS 271-89-6
Molecular Formula C8H6O
Molecular Weight (g/mol) 118.14
MDL Number MFCD00005847
InChI Key IANQTJSKSUMEQM-UHFFFAOYSA-N
Synonym benzofuran, 2,3-benzofuran, coumarone, benzo b furan, cumarone, benzofurfuran, 1-oxindene, coumaron, 1-oxidene
PubChem CID 9223
ChEBI CHEBI:35260
IUPAC Name 1-benzofuran
SMILES O1C=CC2=CC=CC=C12

Specifications

Density 1.095
Boiling Point 173°C to 175°C
Flash Point 56°C (133°F)
Odor Aromatic
Refractive Index 1.566
Quantity 10 g
UN Number UN1993
Beilstein 107704
Merck Index 14,1088
Solubility Information Not miscible in water.
Formula Weight 118.14
Percent Purity ≥99%
Chemical Name or Material Benzo[b]furan
Show More Show Less
Hazard Category H226-H351
Hazard Statement GHS H Statement
H226-H351
Flammable liquid and vapour.
Suspected of causing cancer.
Precautionary Statement P201-P202-P210-P233-P240-P241-P242-P243-P281-P303+P361+P353-P308+P313-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 205-982-6
RTECSNumber DF6423800
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only