accessibility menu, dialog, popup

UserName

CAS RN 271-89-6

IUPAC Name: 1-benzofuran
Synonyms: benzofuran coumaron benzo(b)furan benzofurfuran 1-oxidene cumarone 1-oxindene 2,3-benzofuran
Molecular Weight (g/mol): 118.14
Molecular Formula: C8H6O
InChi Key: IANQTJSKSUMEQM-UHFFFAOYSA-N
SMILES: O1C=CC2=CC=CC=C12
Boiling Point 
  • (4)
  • (2)
Percent Purity 
  • (2)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (2)
  • (4)
  • (4)
  • (2)
  • (2)
  • (2)
  • (2)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (1)
  • (2)
  • (2)