missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-ylacetic acid, 97%, Thermo Scientific™

Catalog No. p-4510630
Change view
Click to view available options
Quantity:
250mg
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
CC40801CB 250mg
CC40801DA 1g
CC40801DE 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC40801CB Supplier Thermo Scientific Chemicals Supplier No. CC40801CB

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 64175-51-5
Molecular Formula C10H8O3
Molecular Weight (g/mol) 176.171
InChI Key QWMVFCMIUUHJDH-UHFFFAOYSA-N
Synonym 2-benzofuran-3-yl acetic acid, 3-benzofuranacetic acid, benzo b furan-3-ylacetic acid, 2-1-benzofuran-3-yl acetic acid, benzofuran-3-yl-acetic acid, benzofuran-3-acetic acid, 1-benzofuran-3-ylacetic acid, 2-3-benzofuranyl acetic acid, pubchem7012, benzofuran-3-ylacetic acid
PubChem CID 2762940
IUPAC Name 2-(1-benzofuran-3-yl)acetic acid
SMILES C1=CC=C2C(=C1)C(=CO2)CC(=O)O

Specifications

Assay Percent Range 97%
Quantity 250mg
Formula Weight 176.17
Percent Purity 97%
Chemical Name or Material Benzo[b]furan-3-ylacetic acid
Hazard Category Warning