accessibility menu, dialog, popup

UserName

CAS RN 64175-51-5

IUPAC Name: 2-(1-benzofuran-3-yl)acetic acid
Synonyms: 1-benzofuran-3-ylacetic acid
Molecular Weight (g/mol): 176.17
Molecular Formula: C10H8O3
InChi Key: QWMVFCMIUUHJDH-UHFFFAOYSA-N
SMILES: OC(=O)CC1=COC2=CC=CC=C12
Quantity 
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (1)
  • (1)
  • (1)