missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[b]furan-3-ylacetic acid, 97%, Thermo Scientific™
Specifications
Specifications
| Assay Percent Range | 97% |
| Packaging | Amber Glass Bottle |
| Quantity | 5g |
| Formula Weight | 176.17 |
| Percent Purity | 97% |
| Chemical Name or Material | Benzo[b]furan-3-ylacetic acid |
Safety and Handling
| Hazard Category | Warning |