missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-3-carbonyl chloride, 97%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals CC27502DA

Quantity 1g
Packaging Amber Glass Bottle

 View more versions of this product

Catalog No. CC27502DA

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

Specifications

Chemical Identifiers

111964-21-7
180.587
YUHWSRVMUQSCNL-UHFFFAOYSA-N
13024720
C1=CC=C2C(=C1)C(=CO2)C(=O)Cl
C9H5ClO2
MFCD11841066
benzo b furan-3-carbonyl chloride, 3-benzofurancarbonylchloride, benzofuran-3-carbonyl chloride, benzofuran-3-carboxylic acid chloride, acmc-1cgrg
1-benzofuran-3-carbonyl chloride

Specifications

Benzo[b]furan-3-carbonyl chloride
96.5
97%
C9H5ClO2
MFCD11841066
YUHWSRVMUQSCNL-UHFFFAOYSA-N
1-benzofuran-3-carbonyl chloride
13024720
97%
111964-21-7
100.0
Amber Glass Bottle
1g
benzo b furan-3-carbonyl chloride, 3-benzofurancarbonylchloride, benzofuran-3-carbonyl chloride, benzofuran-3-carboxylic acid chloride, acmc-1cgrg
C1=CC=C2C(=C1)C(=CO2)C(=O)Cl
180.587
180.59
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

SDS
Documents

Documents