missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific™

Chemical Identifiers

CAS 496-41-3
Molecular Formula C9H6O3
Molecular Weight (g/mol) 162.14
MDL Number MFCD00005848
InChI Key OFFSPAZVIVZPHU-UHFFFAOYSA-N
Synonym benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid
PubChem CID 10331
IUPAC Name 1-benzofuran-2-carboxylic acid
SMILES OC(=O)C1=CC2=CC=CC=C2O1
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC233260010
SDS View Documents
Thermo Scientific Chemicals
233260010
1g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC233260050
SDS View Documents
Thermo Scientific Chemicals
233260050
5g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC233260250
SDS View Documents
Thermo Scientific Chemicals
233260250
25g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

496-41-3
162.14
OFFSPAZVIVZPHU-UHFFFAOYSA-N
10331
OC(=O)C1=CC2=CC=CC=C2O1
C9H6O3
MFCD00005848
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid
1-benzofuran-2-carboxylic acid

Specifications

496-41-3
100.0
98%
C9H6O3
1g
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid
OFFSPAZVIVZPHU-UHFFFAOYSA-N
1-benzofuran-2-carboxylic acid
10331
98%
97.5
Authentic
Glass bottle
MFCD00005848
15, 2549
Solubility in water: partially soluble in water,soluble in boiling wat. Other solubilities: good soluble in acetone and ethanol,slightly soluble in chloroform,carbon disulfide
OC(=O)C1=CC2=CC=CC=C2O1
162.14
162.14
Benzo[b]furan-2-carboxylic acid
Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

EINECSNumber : 207-818-9

SDS
Documents

Documents

RUO – Research Use Only