missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-carboxylic acid, 98%, Thermo Scientific™

Supplier:  Thermo Scientific Chemicals 233260050

 View more versions of this product

Catalog No. AC233260050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description

Description

This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Specifications

Benzo[b]furan-2-carboxylic acid
97.5
Authentic
Glass bottle
MFCD00005848
15, 2549
Solubility in water: partially soluble in water,soluble in boiling wat. Other solubilities: good soluble in acetone and ethanol,slightly soluble in chloroform,carbon disulfide
OC(=O)C1=CC2=CC=CC=C2O1
162.14
162.14
496-41-3
100.0
98%
C9H6O3
5g
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid
OFFSPAZVIVZPHU-UHFFFAOYSA-N
1-benzofuran-2-carboxylic acid
10331
98%

Chemical Identifiers

496-41-3
162.14
OFFSPAZVIVZPHU-UHFFFAOYSA-N
10331
OC(=O)C1=CC2=CC=CC=C2O1
C9H6O3
MFCD00005848
benzofuran-2-carboxylic acid, coumarilic acid, benzo b furan-2-carboxylic acid, 2-benzofurancarboxylic acid, 2-carboxybenzofuran, coumarone-2-carboxylic acid, unii-7z6nmg947l, chembl84095, benzofuran-2-carboxylicacid
1-benzofuran-2-carboxylic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
Causes serious eye irritation.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
Wash face,hands and any exposed skin thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for bre

GHS Signal Word: Warning

EINECSNumber : 207-818-9

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only