Learn More
Benzo[b]furan-2-boronic acid, 98%
CAS: 98437-24-2 | C8H7BO3 | 161.95 g/mol
$94.97 - $854.41
Chemical Identifiers
| CAS | 98437-24-2 |
|---|---|
| Molecular Formula | C8H7BO3 |
| Molecular Weight (g/mol) | 161.95 |
| MDL Number | MFCD00236019 |
| InChI Key | PKRRNTJIHGOMRC-UHFFFAOYSA-N |
| Synonym | benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid |
| PubChem CID | 2776266 |
| IUPAC Name | 1-benzofuran-2-ylboronic acid |
| SMILES | OB(O)C1=CC2=CC=CC=C2O1 |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2367603
|
Thermo Scientific Chemicals
B2367603 |
1 g |
Each for $94.97
|
|
|||||
|
AAB2367606
|
Thermo Scientific Chemicals
B2367606 |
5 g |
Each for $217.14
|
|
|||||
|
AAB2367614
|
Thermo Scientific Chemicals
B2367614 |
25 g |
Each for $854.41
|
|
|||||
Description
It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsIt is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.
Solubility
Soluble in water.
Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
Chemical Identifiers
| 98437-24-2 | |
| 161.95 | |
| PKRRNTJIHGOMRC-UHFFFAOYSA-N | |
| 2776266 | |
| OB(O)C1=CC2=CC=CC=C2O1 |
| C8H7BO3 | |
| MFCD00236019 | |
| benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid | |
| 1-benzofuran-2-ylboronic acid |
Specifications
| 98437-24-2 | |
| MFCD00236019 | |
| 1637889 | |
| Soluble in water. | |
| OB(O)C1=CC2=CC=CC=C2O1 | |
| 161.95 | |
| 161.95 | |
| Benzo[b]furan-2-boronic acid |
| C8H7BO3 | |
| 1 g | |
| benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid | |
| PKRRNTJIHGOMRC-UHFFFAOYSA-N | |
| 1-benzofuran-2-ylboronic acid | |
| 2776266 | |
| 98% |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only