missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]furan-2-boronic acid, 98%

CAS: 98437-24-2 | C8H7BO3 | 161.95 g/mol

Supplier:  Thermo Scientific Chemicals B2367606

 View more versions of this product

Catalog No. AAB2367606


Only null left
Explore available promotions

Description

Description

It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the facile preparation of highly-functionalized, nitrogen-bearing diarylmethanes.

Solubility
Soluble in water.

Notes
Store away from water/moisture and oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Store under inert gas. Protect from humidity and water.
Specifications

Specifications

Benzo[b]furan-2-boronic acid
C8H7BO3
5 g
benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
PKRRNTJIHGOMRC-UHFFFAOYSA-N
1-benzofuran-2-ylboronic acid
2776266
98%
98437-24-2
MFCD00236019
1637889
Soluble in water.
OB(O)C1=CC2=CC=CC=C2O1
161.95
161.95

Chemical Identifiers

98437-24-2
161.95
PKRRNTJIHGOMRC-UHFFFAOYSA-N
2776266
OB(O)C1=CC2=CC=CC=C2O1
C8H7BO3
MFCD00236019
benzofuran-2-boronic acid, benzo b furan-2-boronic acid, benzofuran-2-ylboronic acid, 2-benzofuranboronic acid, 2-benzofuranylboronic acid, 2,3-benzo b furan-2-boronic acid, 2-boronobenzo b furan, 1-benzofuran-2-yl boronic acid, 1-benzofuran-2-boronic acid
1-benzofuran-2-ylboronic acid
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c

H315-H319-H335

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only