missing translation for 'onlineSavingsMsg'
Learn More

Benzo[b]benzo[4,5]thieno[2,3-d]thiophene 97.0+%, TCI America™

Catalog No. B5551200MG
Change view
Click to view available options
Quantity:
200 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B5551200MG 200 mg
B55511G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B5551200MG Supplier TCI America Supplier No. B5551200MG

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 248-70-4
Molecular Formula C14H8S2
Molecular Weight (g/mol) 240.338
MDL Number MFCD02114004
InChI Key NXCSDJOTXUWERI-UHFFFAOYSA-N
PubChem CID 136056
IUPAC Name [1]benzothiolo[3,2-b][1]benzothiole
SMILES C1=CC=C2C(=C1)C3=C(S2)C4=CC=CC=C4S3

Specifications

Melting Point 226°C
Color Yellow
Quantity 200 mg
Formula Weight 240.34
Percent Purity ≥97.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[b]benzo[4,5]thieno[2,3-d]thiophene