accessibility menu, dialog, popup

UserName

CAS RN 248-70-4

IUPAC Name: 8,16-dithiatetracyclo[7.7.0.0²,⁷.0¹⁰,¹⁵]hexadeca-1(9),2,4,6,10(15),11,13-heptaene
Synonyms: 8,16-dithiatetracyclo[7.7.0.0²,⁷.0¹⁰,¹⁵]hexadeca-1(9),2,4,6,10(15),11,13-heptaene
Molecular Weight (g/mol): 240.34
Molecular Formula: C14H8S2
InChi Key: NXCSDJOTXUWERI-UHFFFAOYSA-N
SMILES: S1C2=CC=CC=C2C2=C1C1=C(S2)C=CC=C1
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results