missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo-18-crown 6-Ether 96.0+%, TCI America™
$117.44 - $469.74
Chemical Identifiers
| CAS | 14098-24-9 |
|---|---|
| Molecular Formula | C16H24O6 |
| Molecular Weight (g/mol) | 312.362 |
| MDL Number | MFCD00062741 |
| InChI Key | DSFHXKRFDFROER-UHFFFAOYSA-N |
| PubChem CID | 585779 |
| IUPAC Name | 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(22),18,20-triene |
| SMILES | C1COCCOCCOC2=CC=CC=C2OCCOCCO1 |
Chemical Identifiers
| 14098-24-9 | |
| 312.362 | |
| DSFHXKRFDFROER-UHFFFAOYSA-N | |
| 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(22),18,20-triene |
| C16H24O6 | |
| MFCD00062741 | |
| 585779 | |
| C1COCCOCCOC2=CC=CC=C2OCCOCCO1 |
Specifications
| 14098-24-9 | |
| White | |
| MFCD00062741 | |
| benzo-18-crown-6, b18c6-benzo crown ether, benzo-18-crown 6-ether, 2,3,5,6,8,9,11,12,14,15-decahydrobenzo b 1,4,7,10,13,16 hexaoxacyclooctadecine, 2,3-benzo-1,4,7,10,13,16-hexaoxaoctadec-2-ene, monobenzo-18-crown-6, benzo-18-crown6-ether, 2,3-benzo-18-crown-6, benzo-18-crown, benzo-18-crown 1g | |
| C1COCCOCCOC2=CC=CC=C2OCCOCCO1 | |
| 312.362 | |
| 312.36 | |
| Crystalline Powder |
| 44°C | |
| C16H24O6 | |
| 1 g | |
| DSFHXKRFDFROER-UHFFFAOYSA-N | |
| 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(22),18,20-triene | |
| 585779 | |
| ≥96.0% (GC) | |
| Benzo-18-crown 6-Ether |
Safety and Handling
TSCA : No