accessibility menu, dialog, popup

UserName

CAS RN 14098-24-9

OOOOOO

CAS RN 14098-24-9

IUPAC Name: 2,3,5,6,8,9,11,12,14,15-decahydro-1,4,7,10,13,16-benzohexaoxacyclooctadecine
Synonyms: B18C6-benzo crown ether
Molecular Weight (g/mol): 312.36
Molecular Formula: C16H24O6
InChi Key: DSFHXKRFDFROER-UHFFFAOYSA-N
SMILES: C1COCCOCCOC2=CC=CC=C2OCCOCCO1
Molecular Weight (g/mol) 
  • (2)
  • (5)
Percent Purity 
  • (2)
  • (5)
Quantity 
  • (3)
  • (1)
  • (3)
Physical Form 
  • (2)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (5)
  • (2)
  • (5)
  • (2)
  • (5)
  • (3)
  • (1)
  • (3)
  • (2)
  • (2)