missing translation for 'onlineSavingsMsg'
Learn More

Benzo-18-crown 6-Ether 96.0+%, TCI America™

Catalog No. B15391G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B15391G 1 g
B15395G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B15391G Supplier TCI America Supplier No. B15391G

Chemical Identifiers

CAS 14098-24-9
Molecular Formula C16H24O6
Molecular Weight (g/mol) 312.362
MDL Number MFCD00062741
InChI Key DSFHXKRFDFROER-UHFFFAOYSA-N
PubChem CID 585779
IUPAC Name 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(22),18,20-triene
SMILES C1COCCOCCOC2=CC=CC=C2OCCOCCO1

Specifications

Melting Point 44°C
Color White
Quantity 1 g
Synonym benzo-18-crown-6, b18c6-benzo crown ether, benzo-18-crown 6-ether, 2,3,5,6,8,9,11,12,14,15-decahydrobenzo b 1,4,7,10,13,16 hexaoxacyclooctadecine, 2,3-benzo-1,4,7,10,13,16-hexaoxaoctadec-2-ene, monobenzo-18-crown-6, benzo-18-crown6-ether, 2,3-benzo-18-crown-6, benzo-18-crown, benzo-18-crown 1g
Formula Weight 312.36
Percent Purity ≥96.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo-18-crown 6-Ether