missing translation for 'onlineSavingsMsg'
Learn More

Benzo-18-crown-6, 97%

Catalog No. AAA1622314
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1622314 25 g
AAA1622303 1 g
AAA1622306 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAA1622314 Supplier Thermo Scientific Chemicals Supplier No. A1622314
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 14098-24-9 | C16H24O6 | 312.362 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 14098-24-9
Molecular Formula C16H24O6
Molecular Weight (g/mol) 312.362
MDL Number MFCD00062741
InChI Key DSFHXKRFDFROER-UHFFFAOYSA-N
PubChem CID 585779
IUPAC Name 2,5,8,11,14,17-hexaoxabicyclo[16.4.0]docosa-1(22),18,20-triene
SMILES C1COCCOCCOC2=CC=CC=C2OCCOCCO1

Specifications

Melting Point 43°C to 45°C
Quantity 25 g
Beilstein 1626123
Sensitivity Hygroscopic
Synonym benzo-18-crown-6, b18c6-benzo crown ether, benzo-18-crown 6-ether, 2,3,5,6,8,9,11,12,14,15-decahydrobenzo b 1,4,7,10,13,16 hexaoxacyclooctadecine, 2,3-benzo-1,4,7,10,13,16-hexaoxaoctadec-2-ene, monobenzo-18-crown-6, benzo-18-crown6-ether, 2,3-benzo-18-crown-6, benzo-18-crown, benzo-18-crown 1g
Formula Weight 312.37
Percent Purity 97%
Chemical Name or Material Benzo-18-crown-6
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
TSCA No
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only