missing translation for 'onlineSavingsMsg'
Learn More

Benzo-15-crown 5-Ether 98.0+%, TCI America™

Catalog No. B11541G
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B11541G 1 g
B11545G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B11541G Supplier TCI America Supplier No. B11541G
Only null left

Chemical Identifiers

CAS 14098-44-3
Molecular Formula C14H20O5
Molecular Weight (g/mol) 268.31
MDL Number MFCD00005945
InChI Key FNEPSTUXZLEUCK-UHFFFAOYSA-N
Synonym benzo-15-crown-5, 2,3,5,6,8,9,11,12-octahydrobenzo b 1,4,7,10,13 pentaoxacyclopentadecine, monobenzo-15-crown-5, benzyl 15-crown-5, benzo15c5, denzo15c5, 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine, benzo-15-crown-5-ether, benzo-15-crown 5-ether, ccris 3608
PubChem CID 84197
ChEBI CHEBI:37444
IUPAC Name 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine
SMILES C1COCCOC2=CC=CC=C2OCCOCCO1

Specifications

Melting Point 80°C
Color White
Quantity 1 g
Formula Weight 268.31
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo-15-crown 5-Ether
RTECSNumber DI5400000
TSCA No