accessibility menu, dialog, popup

UserName

OOOOO

CAS RN 14098-44-3

IUPAC Name: 2,3,5,6,8,9,11,12-octahydro-1,4,7,10,13-benzopentaoxacyclopentadecine
Synonyms: benzo-15-crown-5 benzo15C5 denzo15C5 monobenzo-15-crown-5 benzo-15-crown-5-ether benzyl 15-crown-5
Molecular Weight (g/mol): 268.31
Molecular Formula: C14H20O5
InChi Key: FNEPSTUXZLEUCK-UHFFFAOYSA-N
SMILES: C1COCCOC2=CC=CC=C2OCCOCCO1
Percent Purity 
  • (2)
  • (2)
Quantity 
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)
  • (2)