Learn More
Benzo-15-crown-5, 98%
Description
Benzo-15-crown-5 is used in synthetic chemistry. It plays a major role and involved in complexation through ether oxygen. It is used in phase-transfer catalyst system. It is involved in 'Host-Guest' chemistry to identify the move of essential elements in the body. It can play the part of very complicated biological reactions such as enzyme functions, which finds application in the development of new pharmaceuticals.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Benzo-15-crown-5 is used in synthetic chemistry. It plays a major role and involved in complexation through ether oxygen. It is used in phase-transfer catalyst system. It is involved in ′Host-Guest′ chemistry to identify the move of essential elements in the body. It can play the part of very complicated biological reactions such as enzyme functions, which finds application in the development of new pharmaceuticals.
Solubility
Soluble in hot methanol. Slightly soluble in water.
Notes
Hygroscopic. Incompatible with strong oxidizing agents. Store in a cool place.
Specifications
Specifications
| Melting Point | 78°C to 82°C |
| Quantity | 5 g |
| Beilstein | 1624106 |
| Solubility Information | Soluble in hot methanol. Slightly soluble in water. |
| Formula Weight | 268.31 |
| Percent Purity | 98% |
| Chemical Name or Material | Benzo-15-crown-5 |
Safety and Handling
| Hazard Category | H315-H319-H335 |
| Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
| Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
| EINECSNumber | 237-947-6 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only