missing translation for 'onlineSavingsMsg'
Learn More

Benzo[1,2-b:4,5-b']dithiophene 98.0+%, TCI America™

Catalog No. p-7122964
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B34641G 1 g
B34645G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B34641G Supplier TCI America Supplier No. B34641G
Only null left

Chemical Identifiers

CAS 267-65-2
Molecular Formula C10H6S2
Molecular Weight (g/mol) 190.278
InChI Key URMVZUQDPPDABD-UHFFFAOYSA-N
PubChem CID 11106168
IUPAC Name thieno[2,3-f][1]benzothiole
SMILES C1=CSC2=CC3=C(C=C21)SC=C3

Specifications

Melting Point 198°C
Quantity 1 g
Formula Weight 190.28
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[1,2-b:4,5-b′]dithiophene