accessibility menu, dialog, popup

UserName

SS

CAS RN 267-65-2

IUPAC Name: 4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Synonyms: 4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1,3(7),5,8,11-pentaene
Molecular Weight (g/mol): 190.28
Molecular Formula: C10H6S2
InChi Key: URMVZUQDPPDABD-UHFFFAOYSA-N
SMILES: S1C=CC2=CC3=C(C=CS3)C=C12
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results