missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzo[1,2-b:4,5-b']dithiophene 98.0+%, TCI America™
Specifications
Specifications
| Melting Point | 198°C |
| Quantity | 1 g |
| Formula Weight | 190.28 |
| Percent Purity | ≥98.0% (GC) |
| Physical Form | Crystalline Powder |
| Chemical Name or Material | Benzo[1,2-b:4,5-b′]dithiophene |
Safety and Handling
| TSCA | No |