missing translation for 'onlineSavingsMsg'
Learn More

Benzo[1,2-b:4,5-b']dithiophene-4,8-dione 98.0+%, TCI America™

Catalog No. p-7123170
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B37931G 1 g
B37935G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B37931G Supplier TCI America Supplier No. B37931G
Only null left

Chemical Identifiers

CAS 32281-36-0
Molecular Formula C10H4O2S2
Molecular Weight (g/mol) 220.26
InChI Key SIUXRPJYVQQBAF-UHFFFAOYSA-N
PubChem CID 288478
IUPAC Name thieno[2,3-f][1]benzothiole-4,8-dione
SMILES C1=CSC2=C1C(=O)C3=C(C2=O)C=CS3

Specifications

Melting Point 260°C
Quantity 1 g
Formula Weight 220.26
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Benzo[1,2-b:4,5-b′]dithiophene-4,8-dione