accessibility menu, dialog, popup

UserName

OSOS

CAS RN 32281-36-0

IUPAC Name: 4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),3(7),5,11-tetraene-2,8-dione
Synonyms: 4,10-dithiatricyclo[7.3.0.0³,⁷]dodeca-1(9),3(7),5,11-tetraene-2,8-dione
Molecular Weight (g/mol): 220.26
Molecular Formula: C10H4O2S2
InChi Key: SIUXRPJYVQQBAF-UHFFFAOYSA-N
SMILES: O=C1C2=C(C=CS2)C(=O)C2=C1C=CS2
Percent Purity 
  • (2)
Physical Form 
  • (2)
Quantity 
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results