missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Benzil Dioxime 98.0+%, TCI America™
Supplier: TCI America D089125G
Specifications
Benzil Dioxime | |
240°C | |
C14H12N2O2 | |
25 g | |
JJZONEUCDUQVGR-VCFJNTAESA-N | |
(Z)-N-[(2E)-2-(hydroxyimino)-1,2-diphenylethylidene]hydroxylamine | |
5369401 | |
≥98.0% (N) |
23873-81-6 | |
White-Yellow | |
MFCD00002113 | |
diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0 | |
O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1 | |
240.26 | |
240.26 | |
Crystalline Powder |
Chemical Identifiers
23873-81-6 | |
240.26 | |
JJZONEUCDUQVGR-VCFJNTAESA-N | |
5369401 | |
O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1 |
C14H12N2O2 | |
MFCD00002113 | |
diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0 | |
(Z)-N-[(2E)-2-(hydroxyimino)-1,2-diphenylethylidene]hydroxylamine |
Safety and Handling
TSCA : Yes