accessibility menu, dialog, popup

UserName

CAS RN 23873-81-6

IUPAC Name: (Z)-N-[(2E)-2-(hydroxyimino)-1,2-diphenylethylidene]hydroxylamine
Synonyms: (Z)-N-[(2E)-2-(hydroxyimino)-1,2-diphenylethylidene]hydroxylamine
Molecular Weight (g/mol): 240.26
Molecular Formula: C14H12N2O2
InChi Key: JJZONEUCDUQVGR-VCFJNTAESA-N
SMILES: O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1
Percent Purity 
  • (1)
  • (2)
Quantity 
  • (1)
  • (2)
Physical Form 
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)
  • (2)
  • (1)