missing translation for 'onlineSavingsMsg'
Learn More

Benzil dioxime, 98%

Catalog No. p-7080939
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1068714 25 g
AAA1068722 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1068714 Supplier Thermo Scientific Chemicals Supplier No. A1068714

CAS: 23873-81-6 | C14H12N2O2 | 240.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 23873-81-6
Molecular Formula C14H12N2O2
Molecular Weight (g/mol) 240.26
MDL Number MFCD00002113
InChI Key JJZONEUCDUQVGR-VCFJNTAESA-N
Synonym diphenylglyoxime, benzil dioxime, ethanedione, diphenyl-, dioxime, e,e, diphenyl glyoxime, beta-benzil dioxime, gamma-benzil dioxime, n-z-2-nitroso-1,2-diphenylethenyl hydroxylamine, unii-ji58npg8m0, unii-pqn86x6991, ji58npg8m0
PubChem CID 5369401
IUPAC Name N-[(Z)-2-nitroso-1,2-diphenylethenyl]hydroxylamine
SMILES O\N=C(\C(=N/O)\C1=CC=CC=C1)/C1=CC=CC=C1

Specifications

Melting Point ∼238°C (decomposition)
Quantity 25 g
Beilstein 2053615
Merck Index 14,1079
Formula Weight 240.26
Percent Purity 98%
Chemical Name or Material Benzil dioxime
EINECSNumber 245-921-0
RTECSNumber DD1985000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only