missing translation for 'onlineSavingsMsg'
Learn More

(Benzhydrylthio)acetic Acid 98.0+%, TCI America™

Catalog No. B41755G
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B41755G 5 g
B417525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B41755G Supplier TCI America Supplier No. B41755G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 63547-22-8
Molecular Formula C15H13O2S
Molecular Weight (g/mol) 257.33
MDL Number MFCD05262040
InChI Key HTHFEDOFDBZPRX-UHFFFAOYSA-M
Synonym (Diphenylmethylthio)acetic Acid
PubChem CID 2077886
IUPAC Name 2-[(diphenylmethyl)sulfanyl]acetate
SMILES [O-]C(=O)CSC(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 130°C
Color White-Yellow
Quantity 5 g
Formula Weight 258.34
Percent Purity ≥98.0% (GC,T)
Physical Form Crystalline Powder
Chemical Name or Material (Benzhydrylthio)acetic Acid
TSCA No
Recommended Storage Freezer