missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzhydrylamine 97.0+%, TCI America™
$68.97 - $579.71
Chemical Identifiers
| CAS | 91-00-9 |
|---|---|
| Molecular Formula | C13H13N |
| Molecular Weight (g/mol) | 183.254 |
| MDL Number | MFCD00008059 |
| InChI Key | MGHPNCMVUAKAIE-UHFFFAOYSA-N |
| Synonym | aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine |
| PubChem CID | 7036 |
| IUPAC Name | diphenylmethanamine |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
B012425ML
|
TCI America
B012425ML |
25 mL |
Each for $68.97
|
|
|||||
|
B0124100ML
|
TCI America
B0124100ML |
100 mL |
Each for $193.86
|
|
|||||
|
B0124500ML
|
TCI America
B0124500ML |
500 mL |
Each for $579.71
|
|
|||||
Chemical Identifiers
| 91-00-9 | |
| 183.254 | |
| MGHPNCMVUAKAIE-UHFFFAOYSA-N | |
| 7036 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)N |
| C13H13N | |
| MFCD00008059 | |
| aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine | |
| diphenylmethanamine |
Specifications
| 91-00-9 | |
| Yellow | |
| C13H13N | |
| 25 mL | |
| MGHPNCMVUAKAIE-UHFFFAOYSA-N | |
| diphenylmethanamine | |
| 7036 | |
| ≥97.0% (GC,T) | |
| Benzhydrylamine |
| 12°C | |
| 304°C | |
| MFCD00008059 | |
| aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)N | |
| 183.254 | |
| 183.25 | |
| Liquid |
Safety and Handling
RTECSNumber : DA4407300
TSCA : No