missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrylamine 97.0+%, TCI America™

Catalog No. B012425ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B012425ML 25 mL
B0124100ML 100 mL
B0124500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B012425ML Supplier TCI America Supplier No. B012425ML

Chemical Identifiers

CAS 91-00-9
Molecular Formula C13H13N
Molecular Weight (g/mol) 183.254
MDL Number MFCD00008059
InChI Key MGHPNCMVUAKAIE-UHFFFAOYSA-N
Synonym aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine
PubChem CID 7036
IUPAC Name diphenylmethanamine
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)N

Specifications

Melting Point 12°C
Color Yellow
Boiling Point 304°C
Quantity 25 mL
Formula Weight 183.25
Percent Purity ≥97.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzhydrylamine
RTECSNumber DA4407300
TSCA No