Learn More
Benzhydrylamine, 97%
CAS: 91-00-9 | C13H13N | 183.254 g/mol
$133.45 - $360.12
Chemical Identifiers
| CAS | 91-00-9 |
|---|---|
| Molecular Formula | C13H13N |
| Molecular Weight (g/mol) | 183.254 |
| MDL Number | MFCD00008059 |
| InChI Key | MGHPNCMVUAKAIE-UHFFFAOYSA-N |
| Synonym | aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine |
| PubChem CID | 7036 |
| IUPAC Name | diphenylmethanamine |
| SMILES | C1=CC=C(C=C1)C(C2=CC=CC=C2)N |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAB2430314
|
Thermo Scientific Chemicals
B2430314 |
25 g |
Each for $133.45
|
|
|||||
|
AAB2430322
|
Thermo Scientific Chemicals
B2430322 |
100 g |
Each for $360.12
|
|
|||||
Description
Benzhydrylamine is useful for chiral resolution. It acts as an active pharmaceutical ingredient intermediate. It is also employed as a building block for chemical synthesis. Further, it is used in the preparation of furan-2-carboxylic acid benzhydrylamide by reacting with furan-2-carbonyl chloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsBenzhydrylamine is useful for chiral resolution. It acts as an active pharmaceutical ingredient intermediate. It is also employed as a building block for chemical synthesis. Further, it is used in the preparation of furan-2-carboxylic acid benzhydrylamide by reacting with furan-2-carbonyl chloride.
Solubility
Miscible with chloroform, dimethylsulfoxide and methanol. Slightly miscible with water.
Notes
Air sensitive and moisture sensitive. Store in a cool place. Keep the container tightly closed in a dry and well-ventilated place. Incompatible with strong acids and strong oxidizing agents.
Chemical Identifiers
| 91-00-9 | |
| 183.254 | |
| MGHPNCMVUAKAIE-UHFFFAOYSA-N | |
| 7036 | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)N |
| C13H13N | |
| MFCD00008059 | |
| aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine | |
| diphenylmethanamine |
Specifications
| 91-00-9 | |
| 1.064 | |
| >110°C (230°F) | |
| 1.595 | |
| MFCD00008059 | |
| UN2735 | |
| Air Sensitive | |
| aminodiphenylmethane, benzhydrylamine, 1,1-diphenylmethylamine, diphenylmethyl amine, 1,1-diphenylmethanamine, alpha-aminodiphenylmethane, methanamine, 1,1-diphenyl, alpha-phenylbenzylamine, benzhydrylamin, benzhydryl amine | |
| MGHPNCMVUAKAIE-UHFFFAOYSA-N | |
| diphenylmethanamine | |
| 7036 | |
| 97% |
| 11°C to 13°C | |
| 294°C to 296°C | |
| C13H13N | |
| (C6H5)2CHNH2 | |
| 25 g | |
| 776434 | |
| 14,1076 | |
| Miscible with chloroform,dimethylsulfoxide and methanol; Slightly miscible with water. | |
| C1=CC=C(C=C1)C(C2=CC=CC=C2)N | |
| 183.254 | |
| 183.25 | |
| Benzhydrylamine |
Safety and Handling
GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
H302-H315-H319-H335
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: AMINES, LIQUID, CORROSIVE, N.O.S.
EINECSNumber : 202-032-2
RTECSNumber : DA4407300
TSCA : No
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only