missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl Chloride 95.0+%, TCI America™

Catalog No. B107825ML
Change view
Click to view available options
Quantity:
25 mL
500 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B107825ML 25 mL
B1078500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B107825ML Supplier TCI America Supplier No. B107825ML

Chemical Identifiers

CAS 90-99-3
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.681
MDL Number MFCD00000855
InChI Key ZDVDCDLBOLSVGM-UHFFFAOYSA-N
Synonym benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene
PubChem CID 7035
IUPAC Name [chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl

Specifications

Color Yellow
Boiling Point 116°C
Quantity 25 mL
UN Number 3265
Formula Weight 202.68
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Benzhydryl Chloride
EINECSNumber (4)-0097
TSCA Yes