missing translation for 'onlineSavingsMsg'
Learn More

Benzhydryl chloride, 98%

Catalog No. p-7027760
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAB2476414 25 g
AAB2476422 100 g
AAB2476436 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AAB2476414 Supplier Thermo Scientific Chemicals Supplier No. B2476414

CAS: 90-99-3 | C13H11Cl | 202.681 g/mol

Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzhydryl chloride was used as initiator during the controlled radical polymerization of styrene catalyzed by ionic iron complex. It was also used as the starting reagent in the synthesis of trimethylhydroquinone derivatives.

Solubility
Insoluble in water.

Notes
Store in a tightly sealed container. Store away from oxidizing agents, bases.

Chemical Identifiers

CAS 90-99-3
Molecular Formula C13H11Cl
Molecular Weight (g/mol) 202.681
MDL Number MFCD00000855
InChI Key ZDVDCDLBOLSVGM-UHFFFAOYSA-N
Synonym benzhydryl chloride, chlorodiphenylmethane, chloromethylene dibenzene, diphenylchloromethane, diphenylmethyl chloride, benzene, 1,1'-chloromethylene bis, chloro phenyl methyl benzene, methane, chlorodiphenyl, unii-cn9n9ayv4b, 1,1'-chloromethylene bisbenzene
PubChem CID 7035
IUPAC Name [chloro(phenyl)methyl]benzene
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)Cl

Specifications

Melting Point 17°C to 20°C
Density 1.14
Boiling Point 139°C to 140°C (3 mmHg)
Flash Point 160°C (320°F)
Odor Pungent
Refractive Index 1.595
Quantity 25 g
UN Number UN3265
Beilstein 607925
Solubility Information Insoluble in water.
Formula Weight 202.69
Percent Purity 98%
Chemical Name or Material Benzhydryl chloride
Show More Show Less
Hazard Category H290-H314
Hazard Statement GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
Precautionary Statement P234-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
EINECSNumber 202-031-7
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only