missing translation for 'onlineSavingsMsg'
Learn More

Benzhydrol, 99%

Catalog No. AC105391000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105391000 100 g Plastic bottle
AC105395000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105391000 Supplier Thermo Scientific Chemicals Supplier No. 105391000
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 91-01-0 | C13H12O | 184.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 91-01-0
Molecular Formula C13H12O
Molecular Weight (g/mol) 184.24
MDL Number MFCD00004488
InChI Key QILSFLSDHQAZET-UHFFFAOYSA-N
Synonym benzhydrol, diphenylcarbinol, benzohydrol, benzhydryl alcohol, hydroxydiphenylmethane, diphenylmethyl alcohol, diphenyl carbinol, alpha-phenylbenzenemethanol, diphenyl-methanol, benzenemethanol, .alpha.-phenyl
PubChem CID 7037
IUPAC Name diphenylmethanol
SMILES C1=CC=C(C=C1)C(C2=CC=CC=C2)O

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 65.0°C to 68.0°C
Color White to Beige
Boiling Point 297.0°C to 298.0°C
Flash Point 160°C
Loss on Drying 0.5% max. (55°C)
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic bottle
Linear Formula (C6H5)2CHOH
Quantity 100 g
Beilstein 06, IV, 4648
Merck Index 15, 1092
Solubility Information Solubility in water: 0.5g/L (20°C)
Formula Weight 184.24
Percent Purity 99%
Physical Form Crystalline Solid
Chemical Name or Material Benzhydrol, 99%
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.
Precautionary Statement GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap a
EINECSNumber 202-033-8
RTECSNumber DC7452000
TSCA TSCA

RUO – Research Use Only