missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride, USP, 97-103%, Spectrum™ Chemical

Catalog No. 18600886
Change view
Click to view available options
Quantity:
12 kg
Packaging:
Amber Glass Bottle
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18600886 12 kg Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18600886 Supplier Spectrum Chemical Mfg Cor Supplier No. BE12912KGBL
Get Up to $500 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

BE129, 121-54-0, C27H42ClNO2

Spectrum™ Chemical Benzethonium Chloride, USP exhibits a broad spectrum of microbiocidal activity against bacteria, fungi, mold and viruses. All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.09
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
IUPAC Name benzyldimethyl(2-{2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy}ethyl)azanium chloride
SMILES [Cl-].CC(C)(C)CC(C)(C)C1=CC=C(OCCOCC[N+](C)(C)CC2=CC=CC=C2)C=C1

Specifications

CAS Max % 100%
Ignition Residue 0.1%
Loss on Drying 5%
Packaging Amber Glass Bottle
Quantity 12 kg
UN Number UN3261
Percent Purity 97 to 103%
Grade USP
Physical Form Crystalline Powder or Lumps