missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium Chloride 97.0+%, TCI America™

Catalog No. p-7118321
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B004425G 25 g
B0044100G 100 g
B0044500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B004425G Supplier TCI America Supplier No. B004425G
Only null left

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.088
MDL Number MFCD00011742
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
PubChem CID 8478
ChEBI CHEBI:31264
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

Melting Point 163°C
Color White
pH 4.8 to 5.5 (1% H2O soln.)
Quantity 25 g
Formula Weight 448.09
Percent Purity ≥97.0% (HPLC)
Physical Form Crystalline Powder
Chemical Name or Material Benzethonium Chloride
EINECSNumber (9)-0252&(3)-2782&(9)-0849
RTECSNumber BO7175000
TSCA Yes