missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzethonium chloride, 97%
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Suitable for battery materials development.
Applications
Benzethonium chloride is used as a surface disinfectant and a preservative in food processing and pharmaceutical industries. It is involved in the determination of sodium dodecyl sulfate using sulphan blue as indicator from the mixture containing other ingredients like sodium chloride and sodium sulfate. It is an effective cationic detergent due to its positively charged hydrophilic part of quaternary ammonium ion. It is an active ingredient in medicaments, deodorants and mouthwashes.
Solubility
Soluble in water.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents.
Benzethonium chloride is used as a surface disinfectant and a preservative in food processing and pharmaceutical industries. It is involved in the determination of sodium dodecyl sulfate using sulphan blue as indicator from the mixture containing other ingredients like sodium chloride and sodium sulfate. It is an effective cationic detergent due to its positively charged hydrophilic part of quaternary ammonium ion. It is an active ingredient in medicaments, deodorants and mouthwashes.
Solubility
Soluble in water.
Notes
Hygroscopic. Light sensitive. Incompatible with strong oxidizing agents.
Specifications
Specifications
| Melting Point | 157°C to 165°C |
| Odor | Odorless |
| Quantity | 100 g |
| UN Number | UN1759 |
| Beilstein | 3898548 |
| Sensitivity | Hygroscopic |
| Merck Index | 14,1074 |
| Solubility Information | Soluble in water. |
| SMILES | CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-] |
| IUPAC Name | benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride |
| Show More |
Safety and Handling
| Hazard Category | H301-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P301+P310-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P312-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S. |
| EINECSNumber | 204-479-9 |
| RTECSNumber | BO7175000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only