missing translation for 'onlineSavingsMsg'
Learn More

Benzethonium chloride, 97%

Catalog No. AC105381000
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC105381000 100 g Plastic bottle
AC105385000 500 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC105381000 Supplier Thermo Scientific Chemicals Supplier No. 105381000

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Benzethonium chloride, 97%, C27H42ClNO2, CAS Number-121-54-0, benzethoniumchloride, benzethonium chloride, disilyn, hyamine, kylacol, phemeride, phemerol chloride, phemithyn, quatrachlor, solamin, 2.5kg, 96% min. (on dry substance) (Argentometry), 100.0, 96.0, CHEBI:31264, White, 204-479-9, 01,433 | CAS: 121-54-0 | C27H42ClNO2 | 448.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 121-54-0
Molecular Formula C27H42ClNO2
Molecular Weight (g/mol) 448.08
MDL Number MFCD00011742
InChI Key UREZNYTWGJKWBI-UHFFFAOYSA-M
Synonym benzethonium chloride, quatrachlor, hyamine, phemeride, phemerol chloride, benzethoniumchloride, phemithyn, disilyn, kylacol, solamin
PubChem CID 8478
ChEBI CHEBI:31264
IUPAC Name benzyl-dimethyl-[2-[2-[4-(2,4,4-trimethylpentan-2-yl)phenoxy]ethoxy]ethyl]azanium;chloride
SMILES CC(C)(C)CC(C)(C)C1=CC=C(C=C1)OCCOCC[N+](C)(C)CC2=CC=CC=C2.[Cl-]

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 158.0°C to 163.0°C
Color White
Loss on Drying 5% max. (105°C, 4 hrs)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (on dry substance) (Argentometry)
Packaging Plastic bottle
Quantity 100 g
Fieser 01,433
Merck Index 15, 1076
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohol,acetone and chloroform
Formula Weight 448.08
Percent Purity 97%
Physical Form Amorphous or Crystalline Powder
Chemical Name or Material Benzethonium chloride, 97%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Toxic if swallowed.
Causes severe skin burns and eye damage.
Very toxic to aquatic life.
Precautionary Statement GHS P Statement
Avoid release to the environment.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes
EINECSNumber 204-479-9
RTECSNumber BO7175000
TSCA TSCA

RUO – Research Use Only