missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl Hydrazide 98.0+%, TCI America™

Catalog No. B003825G
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B003825G 25 g
B0038100G 100 g
B0038500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. B003825G Supplier TCI America Supplier No. B003825G

Chemical Identifiers

CAS 80-17-1
Molecular Formula C6H8N2O2S
Molecular Weight (g/mol) 172.20
MDL Number MFCD00007583,MFCD08064450
InChI Key VJRITMATACIYAF-UHFFFAOYSA-N
Synonym benzenesulfonyl hydrazide, porofor, benzenesulfonic acid, hydrazide, porofor bsh, benzenesulfohydrazide, phenylsulfonylhydrazine, genitron bsh, celogen bsh, benzenesulfonylhydrazine, phenylsulfohydrazide
PubChem CID 65723
IUPAC Name benzenesulfonohydrazide
SMILES NNS(=O)(=O)C1=CC=CC=C1

Specifications

Melting Point 105°C
Color White
Quantity 25 g
UN Number 3226
Formula Weight 172.20
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Benzenesulfonyl Hydrazide
RTECSNumber DB6888000
TSCA Yes