missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl fluoride, 98%

Catalog No. AAL0468906
Change view
Click to view available options
Quantity:
1 g
5 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAL0468906 5 g
AAL0468903 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAL0468906 Supplier Thermo Scientific Chemicals Supplier No. L0468906
Get Up to $1,800 in Chemicals with Purchase
View all promotions

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

CAS: 368-43-4 | C6H5FO2S | 160.16 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 368-43-4
Molecular Formula C6H5FO2S
Molecular Weight (g/mol) 160.16
MDL Number MFCD00007412
InChI Key IDIPWEYIBKUDNY-UHFFFAOYSA-N
Synonym phenylsulfonyl fluoride, benzenesulphonyl fluoride, benzenesulfonylfluoride, phenylsulfonylfluoride, benzenesulphonylfluoride, fluorophenylsulfone, pubchem14507, acmc-20alq8, wln: wsfr, benzenesulfonyl fluoride
PubChem CID 67779
IUPAC Name benzenesulfonyl fluoride
SMILES FS(=O)(=O)C1=CC=CC=C1

Specifications

Density 1.334
Boiling Point 203°C to 204°C
Flash Point 87°C (188°F)
Refractive Index 1.492
Quantity 5 g
UN Number UN2922
Beilstein 2206835
Sensitivity Moisture sensitive
Formula Weight 160.17
Percent Purity 98%
Chemical Name or Material Benzenesulfonyl fluoride
Show More Show Less
Hazard Category H227-H302+H312+H332-H314
Hazard Statement GHS H Statement
H301-H311-H331-H314-H318-H227
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Combustible liquid.
Precautionary Statement P210-P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
EINECSNumber 206-707-2
RTECSNumber DB8940000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Nitrogen

RUO – Research Use Only