missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl Chloride 99.0+%, TCI America™

Catalog No. B003625G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
B003625G 25 g
B0036500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. B003625G Supplier TCI America Supplier No. B003625G

Chemical Identifiers

CAS 98-09-9
Molecular Formula C6H5ClO2S
Molecular Weight (g/mol) 176.614
MDL Number MFCD00007426
InChI Key CSKNSYBAZOQPLR-UHFFFAOYSA-N
Synonym phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride
PubChem CID 7369
IUPAC Name benzenesulfonyl chloride
SMILES C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Color Yellow
Boiling Point 252°C
Quantity 25 g
UN Number 2225
Formula Weight 176.61
Percent Purity ≥99.0% (GC,T)
Physical Form Liquid
Chemical Name or Material Benzenesulfonyl Chloride
EINECSNumber (3)-1893
RTECSNumber DB8750000
TSCA Yes