accessibility menu, dialog, popup

UserName

CAS RN 98-09-9

IUPAC Name: benzenesulfonyl chloride
Synonyms: benzenesulfonyl chloride
Molecular Weight (g/mol): 176.61
Molecular Formula: C6H5ClO2S
InChi Key: CSKNSYBAZOQPLR-UHFFFAOYSA-N
SMILES: ClS(=O)(=O)C1=CC=CC=C1
Boiling Point 
  • (2)
  • (5)
Molecular Weight (g/mol) 
  • (5)
  • (2)
Percent Purity 
  • (3)
  • (2)
  • (2)
Quantity 
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (4)
  • (3)
  • (4)
  • (2)
  • (5)
  • (5)
  • (2)
  • (3)
  • (2)
  • (2)
  • (1)
  • (1)
  • (2)
  • (1)
  • (1)
  • (1)
  • (5)