missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonyl chloride, 98%

Catalog No. aaa108490b
Change view
Click to view available options
Quantity:
250 g
1000 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA108490B 1000 g
AAA1084930 250 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.
Catalog No. AAA108490B Supplier Thermo Scientific Chemicals Supplier No. A108490B
Only null left
This chemical may require us to obtain additional information for our regulatory and chemical compliance records. If required, we will contact you for this information once your order is placed.

CAS: 98-09-9 | C6H5ClO2S | 176.614 g/mol

Benzenesulfonyl chloride reacts with Grignard reagents to form oxindoles from N-unsubstituted indoles. It is widely used to check the assay of thiamine in different food products. It is involved in the synthesis of alpha-disulfones, sulfonamides and sulfoante esters as precursor. It is a derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry. It is common reagent used in Hinsberg test for detection and distinguishing the type of amines as primary, secondary and tertiary amines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Benzenesulfonyl chloride reacts with Grignard reagents to form oxindoles from N-unsubstituted indoles. It is widely used to check the assay of thiamine in different food products. It is involved in the synthesis of alpha-disulfones, sulfonamides and sulfoante esters as precursor. It is a derivatization reagent for the determination of various amines in waste water and surface water at the sub-ppb level by gas chromatography-mass spectrometry. It is common reagent used in Hinsberg test for detection and distinguishing the type of amines as primary, secondary and tertiary amines.

Solubility
Insoluble in water. Soluble in ether and alcohol.

Notes
Moisture sensitive. Incompatible with water, strong oxidizing agents, strong bases, methyl formamide and dimethyl sulfoxide.

Chemical Identifiers

CAS 98-09-9
Molecular Formula C6H5ClO2S
Molecular Weight (g/mol) 176.614
MDL Number MFCD00007426
InChI Key CSKNSYBAZOQPLR-UHFFFAOYSA-N
Synonym phenylsulfonyl chloride, benzene sulfochloride, benzenesulfonic chloride, benzenosulfochloride, benzene sulfonechloride, benzenosulfochlorek, benzenesulfonic acid chloride, benzenesulphonyl chloride, benzolsulfochloride, benzenesulfochloride
PubChem CID 7369
IUPAC Name benzenesulfonyl chloride
SMILES C1=CC=C(C=C1)S(=O)(=O)Cl

Specifications

Melting Point 14°C to 15°C
Density 1.377
Boiling Point 252°C
Flash Point 132°C (270°F)
Odor Irritating
Refractive Index 1.5525
Quantity 1000 g
UN Number UN2225
Beilstein 606926
Sensitivity Moisture sensitive
Merck Index 14,1072
Solubility Information Insoluble in water. Soluble in ether and alcohol.
Formula Weight 176.62
Percent Purity 98%
Chemical Name or Material Benzenesulfonyl chloride
Show More Show Less
Hazard Category H302-H314
Hazard Statement GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Precautionary Statement P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c
DOTInformation Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: BENZENE SULFONYL CHLORIDE
EINECSNumber 202-636-6
RTECSNumber DB8750000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only