missing translation for 'onlineSavingsMsg'
Learn More

Benzenesulfonic acid, tech. ca 75% w/w aq. soln.

CAS: 98-11-3 | C6H6O3S | 158.171 g/mol

Supplier:  Thermo Scientific Chemicals L190770J

 View more versions of this product

Catalog No. AAL190770J


Only null left
Explore available promotions

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

Benzenesulfonic acid
1.3
C6H6O3S
5000 mL
742513
benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
C1=CC=C(C=C1)S(=O)(=O)O
158.171
CHEBI:64455
Technical
98-11-3
>110°C (230°F)
MFCD00011689
UN2586
14,1070
SRSXLGNVWSONIS-UHFFFAOYSA-N
benzenesulfonic acid
7371
158.18
≈75% w/w aq. soln.

Chemical Identifiers

98-11-3
158.171
SRSXLGNVWSONIS-UHFFFAOYSA-N
7371
benzenesulfonic acid
C6H6O3S
MFCD00011689
benzenesulphonic acid, phenylsulfonic acid, besylic acid, benzene sulphonic acid, benzenemonosulfonic acid, benzene sulfonic acid, kyselina benzensulfonova, benzensulfonic acid, ccris 4595, kyselina benzensulfonova czech
CHEBI:64455
C1=CC=C(C=C1)S(=O)(=O)O
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318-H302
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.

P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID

EINECSNumber : 202-638-7

RTECSNumber : DB4200000

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only