missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Benzenesulfonic acid, tech. ca 75% w/w aq. soln.
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Density | 1.3 |
| Flash Point | >110°C (230°F) |
| Quantity | 250 mL |
| UN Number | UN2586 |
| Beilstein | 742513 |
| Merck Index | 14,1070 |
| Formula Weight | 158.18 |
| Grade | Technical |
| Concentration or Composition (by Analyte or Components) | ≈75% w/w aq. soln. |
| Chemical Name or Material | Benzenesulfonic acid |
Safety and Handling
| Hazard Category | H302-H314-H335 |
| Hazard Statement | GHS H Statement H314-H318-H302 Causes severe skin burns and eye damage. Causes serious eye damage. Harmful if swallowed. |
| Precautionary Statement | P260-P264b-P270-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c |
| DOTInformation | Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: ARYL SULFONIC ACIDS, LIQUID |
| EINECSNumber | 202-638-7 |
| RTECSNumber | DB4200000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only